VWR® ARISTAR®, ICP Multi-Element Solution According USP 232 Parental dose, 6 Elements 10 mg/L in 15% Hydrochloric Acid
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® ARISTAR®
85039.180EA 265 GBP
85039.180
VWR® ARISTAR®, ICP Multi-Element Solution According USP 232 Parental dose, 6 Elements 10 mg/L in 15% Hydrochloric Acid
Standards Cationic Standards Multi-Element Standards
Six elements: 10 mg/L each of Ir; Pt; Os; Rh; Pd; Ru in HCl 15%.
Certifications: USP 232. Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.