Film cleaner, Swift™
Supplier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Swift™
786-678EA 392 GBP
786-678
Film cleaner, Swift™
Film Processing Supplies
Swift™ Film Cleaner allows researchers to clean film that has been overexposed or has a high background or speckling without having to repeat experiments. Suitable for all exposed film developed for gel shift assays, Western, Southern and Northern blots.
  • Save time by eliminating need to repeating experiment
  • Provides even signal removal to preserve results
  • Rapidly stopped once optimal signal achieved