Slide filling cabinet
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RAYLE/43.12EA 0 GBP
RAYLE/43.12
Slide filling cabinet
Slide Holders
Slide filling cabinet designed for storing up to 6000 microscope slides in vertical position.