Protein A, HRP (Horseradish Peroxidase)
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NA9120-1MLEA 472 GBP
GEHE25-0051-72 NA9120-1ML
Protein A, HRP (Horseradish Peroxidase)
Protein Expression Protein Expression Reagents
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species. For use in immunodetection and in situ hybridisation applications.