Water, Ecotainer® endotoxin-free, sterile

Supplier: Local
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Ecotainer®
LOCA0082479EEA 0 GBP
LOCA0082479E
Water, Ecotainer® endotoxin-free, sterile
Water

Rinsing solution in PP bottle.


  • Sterile
  • Pyrogen free

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR