Racks for centrifuge tubes

Supplier: Thomson Instrument
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

THMS932MREA 0 GBP
THMS932MR
Racks for centrifuge tubes
Racks Tube Racks
Metal.

Suitable for 20 ml and 40 ml centrifuge tubes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR