Solvent bottles, glass coated
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Dionex™
DION045901EA 0 GBP
DION045901
Solvent bottles, glass coated
Bottles HPLC Bottles
Borosilicate glass, coated.
The solvent bottle and cap assembly holds organic solvents or aqueous buffers to be used as extraction solvents for ASE® systems. The quick-connect fittings allow for easy assembly of the bottles to the systems.