Centrifuge tubes
Supplier: MEDLINE SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
409922
MEDL409922EA 179 GBP
MEDL409922
Centrifuge tubes
Tubes Centrifuge Tubes
PP, with green PE screw caps.
  • Graduated
  • White labelling area
  • DNase, RNase, endotoxins and metal free