Closed cup flash tester, Pensky-Martens
Supplier: Stanhope Seta
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STAN13661-2EA 11380 GBP
STAN13661-2
Closed cup flash tester, Pensky-Martens
Flashpoint Testers
Pensky-Martens closed cup flash point testers are suitable for mains gas or gas canisters.
  • Electrically heated cup and two speed stirrers
  • Ambient to 370 °C temperature and variable range rate
  • Forced air cooling facility
Delivery information: Supplied with sample cup, lid, and shutter stand and instruction manual.