Cleanroom wipes, MicroFirst®
Supplier: Berkshire
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MicroFirst®
BERKMF0909-6-20EA 0 GBP
BERKMF.1212.20 BERKMF.0909.20 BERKMF0909-6-20
Cleanroom wipes, MicroFirst®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This non woven wipe is composed of a hydroentangled blend of 45% cellulose and 55% polyester. This combination of properties provides MicroFirst® with the level of absorbency, abrasion resistance and chemical compatibility required for controlled environments and applications where both cleanliness and economy are of the most importance.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Apertured for increased absorbency and increased surface texture for improved cleaning efficiency
  • Low extractable levels and particle and fiber counts
  • Very durable with high wet strength
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
Ideal for polishing and cleaning metals, plastics, epoxy surfaces, for general purpose wiping applications and spill control and for applying and removing cleaning and disinfecting solutions.
Suitable for use in Class 100, ISO 5, M3.5 and higher cleanrooms.
Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.