Disinfectant tablets, Actichlor™
Supplier: Ecolab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SHIE3049670EA 157 GBP
SHIE3049670
Disinfectant tablets, Actichlor™
Cleaning Agents Disinfectants
Chlorine disinfectant based on NaDCC (sodium dichloroisocyanurate).
  • For surface disinfection of blood spills, general clinical environment, instruments, sinks and drains
  • Full spectrum of efficacy
  • Simple to use effervescent tablet format (2,5 g tablet)