Cleanroom gloves, nitrile, sterile, Nitrex 420
Supplier: DISTRIBUTOR UK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Nitrex
DIUKGN37-LARGEEA 983 GBP
DIUKGN37-LARGE DIUKGN37-MEDIUM DIUKGN37XXTRALARGE DIUKGN37-SMALL DIUKGN37-XLARGE
Cleanroom gloves, nitrile, sterile, Nitrex 420
Gloves
These nitrile butadiene rubber gloves are ideal for use in clinics, laboratories and cleanroom environments.
  • Sterile, powder-free, smooth finish
  • Chlorination on donning surface
  • Latex-free
  • Sterilised by gamma irradiation (min. 25 kGy)
Certifications: EN 455-1/-2/-3; AQL 1,5
Packaging: 200 pairs per case.