Sliding microtome, HM 450
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-2708EA 0 GBP
720-2708
Sliding microtome, HM 450
Microtomes and Microtome Assemblies Microtomes
This HM 450 includes an automatic fine sectioning and trimming features with selectable specimen retraction.
  • Integrated and removable waste tray
  • Specimen fine orientation on X/Y axes
  • Memory function and graphical display in two different sizes
Suitable for section paraffin and frozen sections in biological, botanical, and materials science applications.