Sliding microtome, HM 430
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-2707EA 0 GBP
720-2707
Sliding microtome, HM 430
Microtomes and Microtome Assemblies Microtomes
Highest precision guarantees safe operation − The roller-guided knife sledge results in smooth running of the knife sledge for comfortable and non-tiring working with highest stability. The elegant housing is streamlined and includes all ergonomic aspects for a comfortable, intuitive working environment.
  • Integrated and removable waste tray
  • Specimen fine orientation on X-Y axes
  • Memory function and graphical display in two different sizes
Suitable for section paraffin and frozen sections in biological, botanical, and materials science applications.