Dry ice maker, portable, model B
Supplier: Stanhope Seta
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STAN70220-3EA 0 GBP
STAN70220-3
Dry ice maker, portable, model B
Ice Makers
Dry ice makers are a safe, efficient and convenient method of producing pellets and blocks in a range of sizes.
  • Rapid production of consistently sized blocks/pellets
  • Connects directly to liquid CO2 syphon type (dip tube) cylinders
Dry ice/liquid mixtures can produce temperatures down to –78 °C. The latent heat of vaporisation of dry ice is 572 kJ/kg (246 bTU per lb).
Delivery information: It is supplied with pressure hose and gauge.