Digital camera, Globecam
Supplier: Medline
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Globecam
MEDL9554.1019EA 834 GBP
MEDL9554.1019
Digital camera, Globecam
Cameras
This eyepiece camera can be inserted into the tube of a microscope.
  • No need of a trinocular head
  • Parfocal digital micro-image with immediate display
  • Clear image reproduction thanks to the CMOS sensor
Suitable for use with biological and stereo microscopes.