Ethyl-trans-(ethoxymethylene)acetate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl trans-3-ethoxyacrylate, Ethyl (E)-3-Ethoxyacrylate

HC-6543-5G HC-6543-25G
COBBHC-6543-5GEA 0 GBP
COBBHC-6543-5G COBBHC-6543-25G
Ethyl-trans-(ethoxymethylene)acetate
Ethyl trans-(ethoxymethylene)acetate
Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
MDL Number: MFCD00009863
CAS Number: 5941-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR