Manganese(II) chloride 0.1 mol/l in aqueous solution, Millipore®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Manganese dichloride

SIAL48795-1ML-FEA 81.5 GBP
SIAL48795-1ML-F
Manganese(II) chloride 0.1 mol/l in aqueous solution, Millipore®
Manganese (II) chloride
Formula: MnCl₂
MW: 125.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR