6,7-Dimethoxy-4-quinazolone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one

AS22610G100 AS22610G25 AS22610G5
ACTIAS22610G100EA 0 GBP
ACTIAS22610G100 ACTIAS22610G25 ACTIAS22610G5
6,7-Dimethoxy-4-quinazolone
6,7-Dimethoxy-4-quinazolone
Formula: C₁₀H₁₀N₂O₃
MW: 206.2 g/mol
MDL Number: MFCD01570147
CAS Number: 13794-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR