1-(4-Aminophenyl)-4-methylpiperazine ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline
M2154-5G
M2154-1G
TCIAM2154-5GEA
0
GBP
TCIAM2154-5G
TCIAM2154-1G
1-(4-Aminophenyl)-4-methylpiperazine ≥98.0% (by GC)
1-(4-Aminophenyl)-4-methylpiperazine
Formula:
C₁₁H₁₇N₃ MW: 191.28 g/mol Boiling Pt: 180°C |
MDL Number:
MFCD00172703 CAS Number: 16153-81-4 UN: 3263 ADR: 8,III |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...