Tributylvinyltin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

AS51608G1 AS51608G25 AS51608G5
ACTIAS51608G1EA 29 GBP
ACTIAS51608G1 ACTIAS51608G25 ACTIAS51608G5
Tributylvinyltin
Tributylvinyltin
Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 73…74 °C (0.1 mmHg)
Density: 1.085 g/cm³ (25 °C)
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR