5-Bromo-3-hydroxy-2-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-methylpyridin-3-ol

AS34527G1 AS34527G5
ACTIAS34527G1EA 0 GBP
ACTIAS34527G1 ACTIAS34527G5
5-Bromo-3-hydroxy-2-picoline
5-Bromo-3-hydroxy-2-picoline
Formula: C₆H₆BrNO
MW: 188.0200 g/mol
MDL Number: MFCD12024425
CAS Number: 91420-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR