6,8-Dichloroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6914-5G HC-6914-1G
COBBHC-6914-5GEA 0 GBP
COBBHC-6914-5G COBBHC-6914-1G
6,8-Dichloroquinoline
6,8-Dichloroquinoline
Formula: C₉H₅Cl₂N
MDL Number: MFCD00040711
CAS Number: 703-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR