2-Bromo-5-fluoro-p-xylene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-4-fluoro-2,5-dimethylbenzene

OT-0611-5G OT-0611-25G OT-0611-1G
COBBOT-0611-5GEA 0 GBP
COBBOT-0611-5G COBBOT-0611-25G COBBOT-0611-1G
2-Bromo-5-fluoro-p-xylene
2-Bromo-5-fluoro-p-xylene
Formula: C₈H₈BrF
MW: 203.0500 g/mol
MDL Number: MFCD09878421
CAS Number: 51760-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR