2-Benzyloxy-1,3-dichlorobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Benzyloxy)-1,3-dichlorobenzene

OT-0672-5G OT-0672-1G
COBBOT-0672-5GEA 0 GBP
COBBOT-0672-5G COBBOT-0672-1G
2-Benzyloxy-1,3-dichlorobenzene
2-Benzyloxy-1,3-dichlorobenzene
Formula: C₁₃H₁₀Cl₂O
MDL Number: MFCD11504947
CAS Number: 21524-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR