2-Chloro-5-isopropoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-6388-1G BB-6388-5G
COBBBB-6388-1GEA 0 GBP
COBBBB-6388-1G COBBBB-6388-5G
2-Chloro-5-isopropoxyphenylboronic acid
2-Chloro-5-isopropoxyphenylboronic acid
Formula: C₉H₁₂BClO₃
MDL Number: MFCD16295073
CAS Number: 1256346-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR