4-(Trimethylsilyl)-3-butyn-1-ol
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Trimethylsilyl-3-butyn-1-ol
SILS19110.5
SILS19110.25
SILNSILS19110.5EA
0
GBP
SILNSILS19110.5
SILNSILS19110.25
4-(Trimethylsilyl)-3-butyn-1-ol
4-(Trimethylsilyl)-3-butyn-1-ol
Formula:
C₇H₁₄OSi MW: 142.27 g/mol |
MDL Number:
MFCD00190217 CAS Number: 2117-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...