4-(Trimethylsilyl)-3-butyn-2-ol

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trimethylsilyl)but-3-yn-2-ol

SILS19115.5 SILS19115.25
SILNSILS19115.5EA 0 GBP
SILNSILS19115.5 SILNSILS19115.25
4-(Trimethylsilyl)-3-butyn-2-ol
4-(Trimethylsilyl)-3-butyn-2-ol
Formula: C₇H₁₄OSi
MW: 142.27 g/mol
MDL Number: MFCD00190213
CAS Number: 6999-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR