4-(2-Bromoethyl)aniline hydrochloride
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBAN-3987-1GEA
0
GBP
COBBAN-3987-1G
4-(2-Bromoethyl)aniline hydrochloride
4-(2-Bromoethyl)aniline hydrochloride
Formula:
C₈H₁₁BrClN |
MDL Number:
MFCD04038116 CAS Number: 26407-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...