Perfluorooctadecanoic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

FLUO002269-5GEA 0 GBP
FLUO002269-5G FLUO002269-25G
Perfluorooctadecanoic acid
Perfluorooctadecanoic acid
Formula: C₁₈HF₃₅O₂
MDL Number: MFCD00198021
CAS Number: 16517-11-6
EINECS: 240-582-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR