(1H-Imidazol-4-yl)methanol

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Hydroxymethyl)imidazole

COBBHI-1346-1GEA 0 GBP
COBBHI-1346-1G COBBHI-1346-25G COBBHI-1346-5G
(1H-Imidazol-4-yl)methanol
(1H-Imidazol-4-yl)methanol
Formula: C₄H₆N₂O
MW: 98.1000 g/mol
MDL Number: MFCD00266718
CAS Number: 822-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR