3-Cyano-4-methylaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Amino-2-methylbenzonitrile

M-Clarity™ quality level = MQ100

SIAL549584-25GEA 118 GBP
SIAL549584-25G
3-Cyano-4-methylaniline, Sigma-Aldrich®
3-Cyano-4-methylaniline
Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD00017608
CAS Number: 50670-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR