6-Aminopyridine-2-carbonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS16240G1
AS16240G5
ACTIAS16240G1EA
0
GBP
ACTIAS16240G1
ACTIAS16240G5
6-Aminopyridine-2-carbonitrile
6-Aminopyridine-2-carbonitrile
Formula:
C₆H₅N₃ |
MDL Number:
MFCD11044252 CAS Number: 370556-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...