D-Phenylalaninol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol

AS24104G100 AS24104G25
ACTIAS24104G100EA 97 GBP
ACTIAS24104G100 ACTIAS24104G25
D-Phenylalaninol
D-Phenylalaninol
Formula: C₉H₁₃NO
MW: 151.2 g/mol
MDL Number: MFCD00064298
CAS Number: 5267-64-1
EINECS: 226-086-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR