4-Amino-3,5-dimethylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethylpyridin-4-amine

AS1368G1 AS1368G5
ACTIAS1368G1EA 129 GBP
ACTIAS1368G1 ACTIAS1368G5
4-Amino-3,5-dimethylpyridine
4-Amino-3,5-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
MDL Number: MFCD03413657
CAS Number: 43078-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR