4-Chloropicolinamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Chloropyridine-2-carboxamide
COBBCA-4125-1GEA
0
GBP
COBBCA-4125-1G
COBBCA-4125-5G
4-Chloropicolinamide
4-Chloropicolinamide
Formula:
C₆H₅ClN₂O MW: 156.57 g/mol |
MDL Number:
MFCD01085339 CAS Number: 99586-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...