4-Chloropicolinamide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloropyridine-2-carboxamide

AS23394G25 AS23394G5
ACTIAS23394G25EA 47 GBP
ACTIAS23394G25 ACTIAS23394G5
4-Chloropicolinamide
4-Chloropicolinamide
Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD01085339
CAS Number: 99586-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR