2-Bromo-5-ethoxybenzotrifluoride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Bromo-4-ethoxy-2-(trifluoromethyl)benzene
OT-0817-5G
OT-0817-1G
COBBOT-0817-5GEA
0
GBP
COBBOT-0817-5G
COBBOT-0817-1G
2-Bromo-5-ethoxybenzotrifluoride
2-Bromo-5-ethoxybenzotrifluoride
Formula:
C₉H₈BrF₃O |
MDL Number:
MFCD08059528 CAS Number: 156605-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...