1-(4-Bromophenyl)cyclopropanecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4729-5G CA-4729-1G
COBBCA-4729-5GEA 0 GBP
COBBCA-4729-5G COBBCA-4729-1G
1-(4-Bromophenyl)cyclopropanecarboxylic acid
1-(4-Bromophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉BrO₂
MW: 241.0800 g/mol
MDL Number: MFCD07374439
CAS Number: 345965-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR