5-Fluoro-4-hydroxyquinazoline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008336-1G
008336-5G
FLUO008336-1GEA
0
GBP
FLUO008336-1G
FLUO008336-5G
5-Fluoro-4-hydroxyquinazoline
5-Fluoroquinazolin-4(1H)-one
Formula:
C₈H₅FN₂O MW: 164.14 g/mol |
MDL Number:
MFCD20542997 CAS Number: 436-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...