5-Chloro-4-quinazolone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chloro-3H-quinazolin-4-one, 5-Chloro-3,4-dihydro-4-oxoquinazoline
AS20009G1
AS20009G25
AS20009G5
ACTIAS20009G1EA
53
GBP
ACTIAS20009G1
ACTIAS20009G25
ACTIAS20009G5
5-Chloro-4-quinazolone
5-Chloro-4-quinazolone
Formula:
C₈H₅ClN₂O MW: 180.5911 g/mol |
MDL Number:
MFCD02676498 CAS Number: 60233-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...