N-Benzyl-5-bromopicolinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Benzyl-5-bromopyridine-2-carboxamide

PY-7207-1G PY-7207-5G
COBBPY-7207-1GEA 0 GBP
COBBPY-7207-1G COBBPY-7207-5G
N-Benzyl-5-bromopicolinamide
N-Benzyl 5-bromopicolinamide
Formula: C₁₃H₁₁BrN₂O
MDL Number: MFCD09801059
CAS Number: 951885-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR