4-(4-Fluorobenzoyl)pyridine ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (4-Fluorophenyl)(pyridin-4-yl)methanone

B22327.03 B22327.06 B22327.14
B22327.03EA 17.5 GBP
B22327.03 B22327.06 B22327.14
4-(4-Fluorobenzoyl)pyridine ≥99%
4-(4-Fluorobenzoyl)pyridine

Purity: 99% CAS No.: 41538-36-7 Molecular Formula: C12H8FNO Formula Weight: 201.2 Melting Point: 85-87° MDL No.: MFCD02683088

Formula: C₁₂H₈FNO
MW: 201.2 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD02683088
CAS Number: 41538-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR