5-Acetyl-2-methoxypyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

597767-1G 597767-5G
SIAL597767-1GEA 40.2 GBP
SIAL597767-1G SIAL597767-5G
5-Acetyl-2-methoxypyridine, Sigma-Aldrich®
5-Acetyl-2-methoxypyridine
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD05664350
CAS Number: 213193-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR