5-Acetyl-2-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25044G25 AS25044G5
ACTIAS25044G25EA 280 GBP
ACTIAS25044G25 ACTIAS25044G5
5-Acetyl-2-methoxypyridine
5-Acetyl-2-methoxypyridine
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD05664350
CAS Number: 213193-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR