2-Acetyl-6-methoxypyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(6-Methoxy-2-pyridinyl)ethanone

M-Clarity™ quality level = MQ100

SIAL662542-5GEA 255 GBP
SIAL662542-5G SIAL662542-1G
2-Acetyl-6-methoxypyridine, Sigma-Aldrich®
2-Acetyl-6-methoxypyridine
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD08276836
CAS Number: 21190-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR