2-Acetyl-6-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(6-Methoxy-2-pyridinyl)ethanone

AS25046G1 AS25046G5
ACTIAS25046G1EA 72 GBP
ACTIAS25046G1 ACTIAS25046G5
2-Acetyl-6-methoxypyridine
2-Acetyl-6-methoxypyridine
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD08276836
CAS Number: 21190-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR