4-(2-Bromo-4-nitrophenoxy)tetrahydro-2H-pyran
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2133-5G
HC-2133-1G
COBBHC-2133-5GEA
0
GBP
COBBHC-2133-5G
COBBHC-2133-1G
4-(2-Bromo-4-nitrophenoxy)tetrahydro-2H-pyran
4-(2-Bromo-4-nitrophenoxy)tetrahydro-2H-pyran
Formula:
C₁₁H₁₂BrNO₄ |
MDL Number:
MFCD11504903 CAS Number: 1072944-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...