Methyl 4-(4-formylphenyl)benzoate

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylateMethyl 4'-formylbiphenyl-4-carboxylate4′-Formylbiphenyl-4-carboxylic acid methyl ester

FLUO020821-1GEA 0 GBP
FLUO020821-1G
Methyl 4-(4-formylphenyl)benzoate
Methyl 4-(4-formylphenyl)benzoate
Formula: C₁₅H₁₂O₃
MW: 240.25 g/mol
MDL Number: MFCD04039123
CAS Number: 70916-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR