1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2'-(Benzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

003137-25G 003137-100G
FLUO003137-25GEA 0 GBP
FLUO003137-25G FLUO003137-100G
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Formula: C₁₂H₆F₁₂O₂
MW: 410.16 g/mol
MDL Number: MFCD00042090
CAS Number: 802-93-7
EINECS: 212-354-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR